Learn More
PD-1 Recombinant Llama Monoclonal Antibody (F3-4C9), Invitrogen™
Llama Recombinant Monoclonal Antibody
Supplier:  Thermo Scientific MA549329
Encompass
Catalog No. PIMA549329
Add to Cart
Description
Description
PD-1 Recombinant Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for ELISA, Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Q15116
Llama
Affinity chromatography
RUO
5133
-20° C, Avoid Freeze/Thaw Cycles
scFv
ELISA, Flow Cytometry
F3-4C9
Unconjugated
Pdcd1
CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2
A recombinant protein corresponding to amino acids 25-167 of human PD-1.
100 μg
Primary
Human
Liquid
Product Suggestions
Product Suggestions