Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD 118057, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Human ether-a-go-go (hERG) KV11.1 potassium channel activator. Displays no major effect on INa, ICa/L, IK1 and IKs currents. Causes resting membrane hyperpolarization and decreases action potential duration in rat myocytes in vitro.
Specifications
Quantity | 10 mg |
Formula Weight | 386.27 |
Percent Purity | >99% |
Chemical Name or Material | PD 118057 |
Safety and Handling
Recommended Storage : Store at -20°C