Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD 118057, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Human ether-a-go-go (hERG) KV11.1 potassium channel activator. Displays no major effect on INa, ICa/L, IK1 and IKs currents. Causes resting membrane hyperpolarization and decreases action potential duration in rat myocytes in vitro.
Specifications
CAS | 2830313674-97-4 |
Quantity | 10 mg |
Formula Weight | 386.27 |
Percent Purity | >99% |
Chemical Name or Material | PD 118057 |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction