Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD 135158, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective, nonpeptide CCK2 receptor antagonist (IC50 values are 2.8 and 1232 nM for CCK2 and CCK1 respectively) that displays negligible affinity at GABAA, benzodiazepine, substance P, neurotensin, opioid, bradykinin and 5-HT3 receptors (IC50 > 10 μM). Exhibits anxiolytic activity in elevated plus maze and social interaction tests and increases food intake in rats.
Specifications
Quantity | 50 mg |
Formula Weight | 616.76 |
Percent Purity | >98% |
Chemical Name or Material | PD 135158 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction