Learn More
PD 153035 hydrochloride, Tocris
EGFR-kinase inhibitor
Manufacturer:  R&D Systems 1037/50
Quantity 50mg
 View more versions of this product
Catalog No. 10-375-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
396.67
PD 153035 hydrochloride
C16H14BrN3O2·HCl
4-[(3-Bromophenyl)amino]-6, 7-dimethoxyquinazoline hydrochloride
Specifications
183322-45-4
>98%
396.67
PD 153035 hydrochloride
50mg
C16H14BrN3O2·HCl
4-[(3-Bromophenyl)amino]-6, 7-dimethoxyquinazoline hydrochloride
Safety and Handling
Recommended Storage : Store at Room Temperature