Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD 166793, Tocris Bioscience™

Catalog Number p-4784249
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
199850-67-4
C17H18BrNO4S
412.298
GJOCABIDMCKCEG-INIZCTEOSA-N
s-2-4'-bromo-1,1'-biphenyl-4-ylsulfonamido-3-methylbutanoic acid, unii-u855l3soxd, u855l3soxd, chembl43152, s-2-4'-bromobiphenyl-4-ylsulfonamido-3-methylbutanoic acid, mmp-2/mmp-3 inhibitor iii, pd166793, s-2-4′-bromo-biphenyl-4-sulfonylamino-3-methylbutyric acid, 2s-2-4'-bromo-1,1'-biphenyl-4-sulfonamido-3-methylbutanoic acid, l-valine, n-4'-bromo 1,1'-biphenyl-4-yl sulfonyl, alpha-arylsulfonylamino carboxylate 2s
9887870
(2S)-2-[[4-(4-bromophenyl)phenyl]sulfonylamino]-3-methylbutanoic acid
CC(C)C(C(=O)O)NS(=O)(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)Br
This item is not returnable. View return policy
199850-67-4
C17H18BrNO4S
412.298
GJOCABIDMCKCEG-INIZCTEOSA-N
s-2-4'-bromo-1,1'-biphenyl-4-ylsulfonamido-3-methylbutanoic acid, unii-u855l3soxd, u855l3soxd, chembl43152, s-2-4'-bromobiphenyl-4-ylsulfonamido-3-methylbutanoic acid, mmp-2/mmp-3 inhibitor iii, pd166793, s-2-4′-bromo-biphenyl-4-sulfonylamino-3-methylbutyric acid, 2s-2-4'-bromo-1,1'-biphenyl-4-sulfonamido-3-methylbutanoic acid, l-valine, n-4'-bromo 1,1'-biphenyl-4-yl sulfonyl, alpha-arylsulfonylamino carboxylate 2s
9887870
(2S)-2-[[4-(4-bromophenyl)phenyl]sulfonylamino]-3-methylbutanoic acid
CC(C)C(C(=O)O)NS(=O)(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)Br

Broad spectrum MMP inhibitor

Broad spectrum MMP inhibitor. Displays high affinity for MMP-2, -3 and -13 (IC50 values are 4, 7 and 8 nM respectively) and exhibits > 750-fold selectivity over MMP-1, -7 and -9. Attenuates left ventricular remodelling and dysfunction in rat model of heart failure.
Formula Weight 412.3
Percent Purity >99%
Quantity 10 mg
Chemical Name or Material PD 166793
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit