Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Mouse anti-Human, Mouse, Rat, Clone: P-GF.44C, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | PD-ECGF/Thymidine Phosphorylase |
---|---|
Clone | P-GF.44C |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB253395200
|
Novus Biologicals
NBP253395200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal antibody specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin, SDS-Page.Specifications
PD-ECGF/Thymidine Phosphorylase | |
0.2 mg/mL | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), SDS-Page | |
Unconjugated | |
Mouse | |
Angiogenesis | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
1890 | |
Recombinant full-length human Thymidine Phosphorylase (TP/ PD-ECGF) protein | |
Primary | |
Store at 4C. |
P-GF.44C | |
Western Blot 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase | |
TYMP | |
IgG1 | |
Protein A or G purified | |
55 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title