Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Mouse anti-Human, Mouse, Rat, Clone: rTYMP/3444, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30727010UG
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
PD-ECGF/Thymidine Phosphorylase | |
Monoclonal | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry-Paraffin 1-2 ug/ml | |
EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase | |
Recombinant full-length human PD-ECGF/Thymidine Phosphorylase (TP/ PD-ECGF) protein | |
100 μg | |
Angiogenesis | |
1890 | |
Store at 4°C. | |
IgG1 κ |
Western Blot, Immunohistochemistry (Paraffin) | |
rTYMP/3444 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title