Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-ECGF/Thymidine Phosphorylase Mouse anti-Human, Mouse, Rat, Clone: rTYMP/3444, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen PD-ECGF/Thymidine Phosphorylase
Clone rTYMP/3444
Concentration 0.2 mg/ml
Dilution Western Blot, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122014
View Documents
Novus Biologicals
NBP30727020UG
20 μg
Each for $159.82
Add to cart
 
NB122013
View Documents
Novus Biologicals
NBP30727010UG
100 μg
Each for $374.34
Add to cart
 
Description

Description

PD-ECGF/Thymidine Phosphorylase Monoclonal antibody specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

PD-ECGF/Thymidine Phosphorylase
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Purified
RUO
Human, Mouse, Rat
EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase
Recombinant full-length human PD-ECGF/Thymidine Phosphorylase (TP/ PD-ECGF) protein
Monoclonal
Protein A or G purified
rTYMP/3444
Western Blot, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Angiogenesis
10 mM PBS with 0.05% BSA
1890
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit