Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Rabbit anti-Human, Mouse, Rat, Clone: TYMP/2890R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | PD-ECGF/Thymidine Phosphorylase |
---|---|
Clone | TYMP/2890R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122036
|
Novus Biologicals
NBP30728120UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Immunohistochemistry-Paraffin.Specifications
PD-ECGF/Thymidine Phosphorylase | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Angiogenesis | |
10 mM PBS with 0.05% BSA | |
1890 | |
IgG | |
Protein A or G purified |
TYMP/2890R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase | |
Recombinant full-length human PD-ECGF/Thymidine Phosphorylase (TP/ PD-ECGF) protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title