Learn More
Medchemexpress LLC PD-L1 Human HEK293 50ug
SDP
Supplier:  Medchemexpress LLC HYP7336150UG
Encompass_Preferred
The PD-L1 protein critically regulates immune tolerance by acting as a ligand for PDCD1/PD-1 regulating T cell activation threshold and limiting effector responses It may act as a costimulatory molecule for IL10-producing T cell subsets PD-L1 Protein Human (HEK293) is the recombinant human-derived PD-L1 protein expressed by HEK293 with tag free
Catalog No. 50-001-79134