Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1, Mouse anti-Human,Mouse, Clone: PDL1/2746, Novus Biologicals™

Mouse Monoclonal Antibody

$149.00 - $363.51

Specifications

Antigen PD-L1
Clone PDL1/2746
Applications Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunofluorescence,Immunohistochemistry (Paraffin)
Host Species Mouse
Target Species Human,Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013562
View Documents
Novus Biologicals
NBP280490100UG
100 ug
Each for $363.51
Add to cart
 
NB013563
View Documents
Novus Biologicals
NBP28049020UG
20 ug
Each for $149.00
Add to cart
 
Description

Description

PD-L1 Monoclonal antibody specifically detects PD-L1 in Human, Mouse Samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array.
Specifications

Specifications

PD-L1
Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunofluorescence,Immunohistochemistry (Paraffin)
Human,Mouse
Primary
PDL1/2746
Mouse
Monoclonal
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit