Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L1, Mouse anti-Human,Mouse, Clone: PDL1/2746, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP28049020UG
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Human, Mouse Samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array.Specifications
PD-L1 | |
PDL1/2746 | |
Unconjugated | |
Purified | |
anti-PD-L1, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1 blocking, PD-L1 ihc, PD-L1 immunohistochemistry, PD-L1 western blot, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
CD274 | |
Recombinant fragment (around aa 39-191) of human PD-L1 (PDL1/2746) protein (exact sequence is proprietary) | |
Monoclonal | |
Protein A or G purified | |
RUO | |
Human,Mouse |
Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunofluorescence,Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Protein Array | |
10 mM PBS with 0.05% BSA | |
29126 | |
Mouse | |
IgG2b Kappa | |
Primary | |
20 ug | |
Store at 4°C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title