Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 Rat anti-Human, Mouse, DyLight™ 550, Clone: MIH5, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143262R

Catalog No. NBP143262R


Add to cart

Description

Description

PD-L1 Monoclonal antibody specifically detects PD-L1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

PD-L1
MIH5
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Conjugated, Purified
CD274
The immunogen for this antibody was B7H1.
Protein A or G purified
Signal Transduction
Primary
29126.0
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Rat
IgG2a Lambda
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit