Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L2/B7-DC/PDCD1LG2 Mouse anti-Human, Alexa Fluor 594, Clone: Z64P2D3&asterisk;H4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252408AF594
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PD-L2/B7-DC/PDCD1LG2 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PDCD1LG2 | |
Ovalbumin-conjugated synthetic peptide KAVFFKRHN | |
0.1 mL | |
80380.0 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Z64P2D3*H4 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
Mouse | |
Protein G purified | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title