Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L2 (CD273) Mouse anti-Human, PE, Clone: IPI683.rMab (also known as IPI683.rMAb), BD Pharmingen™
Mouse Monoclonal Antibody
$125.00 - $260.00
Specifications
Antigen | PD-L2 (CD273) |
---|---|
Clone | IPI683.rMab (also known as IPI683.rMAb) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
PD-L2 (CD273) Monoclonal antibody specifically detects PD-L2 (CD273) in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
PD-L2 (CD273) | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
CD273; PD-L2; PDL2; PDCD1LG2; B7-DC; B7DC | |
Human PD-L2 cDNA | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
IPI683.rMab (also known as IPI683.rMAb) | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
80380 | |
IgG1 λ1 | |
Affinity Chromatography |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title