Learn More
PD-L2 (PDCD1LG2) Mouse anti-Human, Clone: OTI12C7, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808982
Catalog No. 50-166-9207
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD273 (PDCD1 Ligand 2, Programmed cell death 2 ligand 2, PD-L2, B7-DC) is involved in cell-mediated immune responses initiated by T lymphocytes that are stimulated by cognate peptides bound to MHC molecules on antigen-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as CD273 that mediate inhibitory signals from the APC (antigen presenting cell). CD273 can bind two different but related ligands, PDL-1 and PDL-2, both of which are thought act as a negative regulator of T cell activation. However, it has been suggested that PDL-2 can act to stimulate an immunogenic response through and alternative receptor from PD-1. At least three isoforms of PDL-2 are known to exist; CD273 is specific to the largest isoform. Diseases associated with CD273 include cysticercosis, and myasthenic syndrome.
Specifications
Specifications
PD-L2 (PDCD1LG2)
Monoclonal
Unconjugated
PDCD1LG2
Lyophilized
PDCD1LG2
Full length human recombit protein of human PDCD1LG2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI12C7
PBS with 8% trehalose and no preservative; pH 7.3
Q9BQ51
B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
80380