Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare PD MidiTrap™ G-25 Sample Preparation Columns : PROMOTION

Promotion Details: Receive a 15% discount.

Convenient, prepacked column for cleanup of biomolecules with a molecular weight (>5000 Mr in sample volumes up to 1mL

Manufacturer:  GE Healthcare 29334692

Catalog No. 45-001-530PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Developed for a wide range of applications: desalting, buffer exchange, removal of low-molecular weight compounds, etc.
  • Equally suited for gravity flow or centrifugation protocols
  • Accommodates sample volumes of 0.5 to 1.0mL for gravity, 0.75 to 1.0mL with centrifugation
  • High recoveries of 70 to 90%, depending on biomolecule
  • High desalting capacityof >90%

Specifications

Specifications

8.5mL
13mm
Each
1.02 in.
>100cm/hr.
Polyethylene
50μm in dry form, 38 to 235μm in wet form
4° to 30°C
0.51 in.
PD MidiTrap Sephadex G-25 gravity column
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel
26mm
Four centrifuge adapters, instructions
2 to 13
Cross-linked Dextran
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit