Learn More
Selleck Chemical LLCSupplier Diversity Partner PD173074 10mg 219580-11-7  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S126410MG
PD173074 is a potent FGFR1 inhibitor with IC50 of ~25 nM and also inhibits VEGFR2 with IC50 of 100-200 nM in cell-free assays, ~1000-fold selective for FGFR1 than PDGFR and c-Src. PD173074 reduces proliferation and promotes apoptosis in gastric cancer cells.
Catalog No. 50-691-2