Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PDAP1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15523820
![]() |
Novus Biologicals
NBP15523820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155238
![]() |
Novus Biologicals
NBP155238 |
100 μL |
Each for $487.50
|
|
|||||
Description
PDAP1 Polyclonal specifically detects PDAP1 in Human samples. It is validated for Western Blot.Specifications
PDAP1 | |
Polyclonal | |
Rabbit | |
Q13442 | |
11333 | |
Synthetic peptides corresponding to PDAP1(PDGFA associated protein 1) The peptide sequence was selected from the N terminal of PDAP1. Peptide sequence MPKGGRKGGHKGRARQYTSPEEIDAQLQAEKQKAREEEEQKEGGDGAAGD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
HASPP28, PAP1PDGFA-associated protein 1, PAP28 kDa heat- and acid-stable phosphoprotein, PDGFA associated protein 1, PDGF-associated protein | |
PDAP1 | |
IgG | |
20 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title