Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD1 Mouse anti-Human, Clone: PDCD1/922, Abnova™

Mouse monoclonal antibody raised against full length recombinant human PDCD1.

Manufacturer:  Abnova Corporation MAB13022

Catalog No. 89-934-745


Add to cart

Description

Description

This gene encodes a cell surface membrane protein of the immunoglobulin superfamily. This protein is expressed in pro-B-cells and is thought to play a role in their differentiation. In mice, expression of this gene is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure. These studies suggest that this gene product may also be important in T cell function and contribute to the prevention of autoimmune diseases. [provided by RefSeq]

Specifications

Specifications

PDCD1
PDCD1/922
Mouse monoclonal antibody raised against full length recombinant human PDCD1.
In PBS (0.05% BSA, 0.05% sodium azide).
CD279/PD1/SLEB2/hPD-1/hPD-l
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
PDCD1
Recombinant protein corresponding to full length human PDCD1.
Protein A/G purification
RUO
Primary
5133
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit