Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD6 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549370

Catalog No. PA549370

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

PDCD6 is a calcium-binding protein required for T cell receptor-, FAS-, and glucocorticoid-induced cell death. It may mediate CA(2+)- regulated signals along the death pathway.
Specifications

Specifications

PDCD6
Polyclonal
PBS with 0.09% sodium azide
P12815
Programmed cell death protein 6, Apoptosis-linked gene 2 protein, Probable calcium-binding protein ALG-2, PDCD6, ALG2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18570
Western Blot
Unconjugated
Pdcd6
Liquid
PDCD6
KLH conjugated synthetic peptide between 103-137 amino acids from the Central region of human PDCD6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit