Learn More
Aviva Systems Biology Corp PDCD7 Antibody - middle region (ARP58777_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58777_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PDCD7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PDCD7. Alternate names: ES18, HES18, MGC22015ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62422