Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCL3 Mouse anti-Human, Mouse, Clone: OTI10A6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527211

Catalog No. PIMA527211


Add to cart

Description

Description

Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL3 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. Along with the related protein PDCL2, PDCL3 interacts with the chaperonin CCT and modulates CCT-mediated actin and tubulin folding. Modulation of PDCL3 levels by MAPK phosphorylation and RhoA-dependent changes also promote cytoskeletal remodeling.
Specifications

Specifications

PDCL3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H2J4, Q8BVF2
VIAF, PHLP3, VIAF1, HTPHLP, PHLP2A, PhLP2A, Phosducin-like protein 3, VIAF-1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
68833, 79031
Immunohistochemistry (Paraffin), Western Blot
OTI10A6
Unconjugated
PDCL3
Liquid
PDCL3
Full length human recombit protein of PDCL3 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit