Learn More
PDE1A Mouse anti-Human, Clone: OTI5C9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803136
Catalog No. 50-166-6342
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cyclic nucleotide phosphodiesterases play a role in signal transduction by regulating intracellular cyclic nucleotide concentrations through hydrolysis of cAMP and/or cGMP to their respective nucleoside 5-prime monophosphates. Members of the PDE1 family, such as PDE1A, are Cathat are activated by calmodulin in the presence of Ca.
Specifications
Specifications
PDE1A
Monoclonal
Unconjugated
PDE1A
Lyophilized
PDE1A
Human recombit protein fragment corresponding to amino acids 259-545 of human PDE1A produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5C9
PBS with 8% trehalose and no preservative; pH 7.3
P54750
CAM-PDE-1A, HCAM-1, HCAM1, HSPDE1A
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5136