Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE1A Mouse anti-Human, Clone: OTI7D5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA803629
Description
Cyclic nucleotide phosphodiesterases play a role in signal transduction by regulating intracellular cyclic nucleotide concentrations through hydrolysis of cAMP and/or cGMP to their respective nucleoside 5-prime monophosphates. Members of the PDE1 family, such as PDE1A, are Cathat are activated by calmodulin in the presence of Ca.Specifications
PDE1A | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P54750 | |
PDE1A | |
Human recombit protein fragment corresponding to amino acids 259-545 of human PDE1A produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7D5 | |
Unconjugated | |
PDE1A | |
CAM-PDE-1A, HCAM-1, HCAM1, HSPDE1A | |
Mouse | |
Affinity Chromatography | |
RUO | |
5136 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction