Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE1B Mouse anti-Human, Clone: OTI6E4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525646

Catalog No. PIMA525646


Add to cart

Description

Description

The protein encoded by this gene belongs to the cyclic nucleotide phosphodiesterase (PDE) family, and PDE1 subfamily. Members of the PDE1 family are calmodulin-dependent PDEs that are stimulated by a calcium-calmodulin complex. This PDE has dual-specificity for the second messengers, cAMP and cGMP, with a preference for cGMP as a substrate. cAMP and cGMP function as key regulators of many important physiological processes. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications

Specifications

PDE1B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q01064
5′′-cyclic nucleotide phosphodiesterase 1B; 63 a Cam PDE; 63 a Cam-PDE; Calcium/calmodulin dependent 3′ 5′ cyclic nucleotide Phosphodiesterase 1B; Calcium/calmodulin stimulated cyclic nucleotide phosphodiesterase; Calcium/calmodulin-dependent 3′′; Calcium/calmodulin-dependent 3′5′-cyclic nucleotide Phosphodiesterase 1B; Calmodulin stimulated phosphodiesterase PDE1B1; Cam PDE 1B; Cam PDE1B; Cam-PDE 1B; PDE 1B; PDE1B; PDE1B; PDE1B1; PDES 1B; PDES1B; Phosphodiesterase 1B; Phosphodiesterase
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5153
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI6E4
Unconjugated
PDE1B
Liquid
PDE1B
Full length human recombit protein of PDE1B produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit