Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE2A Mouse anti-Human, Clone: OTI2A5, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502349

Catalog No. 50-167-3411


Add to cart

Description

Description

PDE2A (phosphodiesterase 2A) is a 100 kD cyclic nucleotide phosphodiesterase (PDE) and three isoforms of PDE2A have been reported (PDE2A1, PDE2A2 and PDE2A3) which arise from a single gene by alternative splicing events. PDE2A can degrade both cAMP and cGMP, but degrades cGMP with greater efficacy. PDE2A is expressed in a variety of tissue types, with greatest expression in the brain, where it is localized in the cerebellum and neocortex. PDE2A has been implicated in penile erectile dysfunction and in the regulation of fluid and inflammatory cell transport across the endothelial cell barrier.
Specifications

Specifications

PDE2A
Monoclonal
0.72 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00408
CGS-PDE, PDE2A1, PED2A4, cGSPDE
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5138
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2A5
Unconjugated
PDE2A
Liquid
PDE2A
Full length human recombit protein of human PDE2A produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit