Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE4A Mouse anti-Human, Clone: OTI7D11, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA501199
Description
Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. PDE4A is a class IV cAMP-specific PDE.Specifications
PDE4A | |
Monoclonal | |
0.7 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P27815 | |
DPDE2, PDE4, PDE46 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
5141 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI7D11 | |
Unconjugated | |
PDE4A | |
Liquid | |
PDE4A | |
Full length human recombit protein of human PDE4A produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title