Learn More
PDE4C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 217541AP
Catalog No. 50-173-3582
Add to cart
Description
Description
Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases.
Specifications
Specifications
PDE4C
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P14644, Q08493, Q3UEI1
DPDE1, PDE21, PDE4C
Rabbit
IgG
150 μL
-20°C
Primary
110385, 290646, 5143
Western Blot
Unconjugated
PDE4C
Liquid
Pde4c
PDE4C Fusion Protein Ag16130
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat