Learn More
PDE4D Mouse anti-Human, Unconjugated, Clone: 1C4B9, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 670621IG150UL
Description
The PDE4D antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PDE4D. This antibody recognizes human, mouse, rat, pig antigen. The PDE4D antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
PDE4D | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, ELISA | |
1C4B9 | |
Unconjugated | |
150 μL | |
5144 | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.