Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE6H Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549248

Catalog No. PA549248


Add to cart

Description

Description

Predicted to react with bovine, rat and human based on sequence homology.

Participates in processes of transmission and amplification of the visual signal. cGMP-PDEs are the effector molecules in G-protein-mediated phototransduction in vertebrate rods and cones.
Specifications

Specifications

PDE6H
Polyclonal
PBS with 0.09% sodium azide
P61249
Retinal cone rhodopsin-sensitive cGMP 3′, 5′-cyclic phosphodiesterase subunit gamma, GMP-PDE gamma, PDE6H
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
78600
Western Blot
Unconjugated
Pde6h
Liquid
PDE6H
KLH conjugated synthetic peptide between 4-33 amino acids from the Central region of human PDE6H.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit