Learn More
PDE8A Mouse anti-Human, DyLight™ 594, Clone: EM-52, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP262213DL594
Catalog No. NB002570
Add to cart
Description
Description
PDE8A Monoclonal specifically detects PDE8A in Human samples. It is validated for Western Blot.
Specifications
Specifications
PDE8A
EM-52
Western Blot
Purified, Conjugated
PDE8A
Human PDE8a
Protein A purified
Lipid and Metabolism, Signal Transduction
Primary
5151
Human
Western Blot
DyLight 594
50mM Sodium Borate with 0.05% Sodium Azide
cAMP-specific cyclic nucleotide phosphodiesterase 8A, EC 3.1.4.17, FLJ16150, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, high-affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, HsT19550, phosphodiesterase 8A
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
The antibody MEM-115 recognizes an epitope in the D1 domain of CD4 antigen, a 55 kDa transmebrane glycoprotein expressed on a subset of T lymphocytes and also on monocytes, tissue macrophages and granulocytes. It is negative in Western blotting even with non-reduced samples of cell lysates.