Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDF Mouse anti-Human, Clone: OTI3F3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502988

Catalog No. 50-167-3977


Add to cart

Description

Description

Removes the formyl group from the N-terminal Met of newly synthesized proteins.
Specifications

Specifications

PDF
Monoclonal
0.78 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9HBH1
peptide deformylase, mitochondrial
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64146
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3F3
Unconjugated
PDF
Liquid
PDF
Full length human recombit protein of human PDF produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit