Learn More
MilliporeSigma™ PDGF (101-116), Rabbit, Unlabeled, Polyclonal,
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ PC21100UG
Catalog No. PC21100UG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Recognizes the B subunit of PDGF in mesenchymal cells and fetal epithelial tissue.
Specifications
Specifications
PDGF (101-116)
Please refer to vial label for lot-specific concentration
In 0.05M sodium phosphate buffer, 0.2% gelatin, ≤0.1% Sodium Azide.
Anti-Platelet Derived Growth Factor
a synthetic peptide (TEVFEISRRLIDRTNA) corresponding amino acids 101-116 of human PDGF B chain
100μg
2°-8°C
Polyclonal
Immunofluorescence
Unconjugated
Liquid
Rabbit
IgG
RUO
Primary
Human, Rat
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.