Learn More
PDGF-AA Goat anti-Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Manufacturer:  R&D Systems AF1055SP
Catalog No. AF1055SP
Add to cart
This item is not returnable. View return policy
Description
Description
PDGF-AA Polyclonal specifically detects PDGF-AA in Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications
Specifications
PDGF-AA
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.2-0.6 ug/mL
AAB59693
Pdgfa
E. coli-derived recombinant rat PDGF-AA Ser87-Arg196 Accession # AAB59693
Immunogen affinity purified
RUO
Primary
5154
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects rat PDGF-AA in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant rat (rr) PDGF-AB is observed, 30% cross-reactivity with recombinant human PDGF-AA is observed, and less than 5% cross-reactivity with rrPDGF-BB is observed.
Rat