Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF-B Mouse anti-Human, Alexa Fluor 405, Clone: MM0014-5F66, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060980AF405
Description
PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PDGF-B | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PDGFB | |
Human recombinant PDGF-B | |
0.1 mL | |
Primary | |
This antibody shows approximately some degree (40%) cross-reactivity with recombinant PDGF-AB and no cross-reactivity with recombinant PDGF-AA. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0014-5F66 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV | |
Mouse | |
Protein G purified | |
Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers | |
5155 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title