Learn More
PDGF-B Mouse anti-Human, DyLight 650, Clone: MM0014-5F66, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11060980C
Catalog No. NB11060980C
Add to cart
Description
Description
PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
PDGF-B
MM0014-5F66
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
PDGFB
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human PDGF BB recombinant protein.
Protein G purified
Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers
Primary
5155
Human
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody shows approximately some degree (40%) cross-reactivity with rPDGF-AB and no cross-reactivity with recombinant PDGF-AA.