Learn More
PDGF-C Mouse anti-Human, Alexa Fluor 488, Clone: 5F12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP221956AF488
Catalog No. NB221956488
Add to cart
Description
Description
PDGF-C Monoclonal specifically detects PDGF-C in Human samples. It is validated for Western Blot.
Specifications
Specifications
PDGF-C
5F12
Western Blot
Purified
PDGFC
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant PDGF-C. Mouse anti human PDGF-C Q9NRA1.
Protein G purified
Cell Cycle and Replication
Primary
56034
Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Human