Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals PDGF R beta Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
Manufacturer: Novus Biologicals NBP2794815PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human PDGF R beta.Specifications
62.5 pg/mL | |
Sandwich ELISA | |
RUO | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human PDGF R beta. | |
PDGF R beta | |
Oncogenes, Phospho Specific, Protein Kinase | |
Human | |
PDGFRB | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to a working concentration of 0.8 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended |
HRP | |
5159 | |
Antibody Pairs | |
62.5-4000 pg/mL | |
Storage is content dependent. | |
Mouse | |
5 Plates | |
Sandwich ELISA | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title