Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Mouse anti-Human, Allophycocyanin/Cy7, Clone: 18A2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP147232APCCY7
Description
PDGF R beta Monoclonal antibody specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry.Specifications
PDGF R beta | |
18A2 | |
Flow Cytometry | |
Conjugated, Purified | |
PDGFRB | |
CD140b-transfected NIH 3T3 cells | |
Protein A purified | |
Oncogenes, Phospho Specific, Protein Kinase | |
Primary | |
5159.0 | |
Human |
Flow Cytometry | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
The monoclonal antibody 18A2 recognizes CD140b/ PDGF-RB, the 180-190 kDa beta chain of platelet-derived growth factor receptor, which is widely expressed on a variety of mesenchymal-derived cells and plays pro-proliferative roles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title