Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Antibody (42G12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP1191910.025MG
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Proximity Ligation Assay, Knockdown Validated.Specifications
PDGF R beta | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
PDGFRB | |
Recombinant human PDGF Receptor beta Protein | |
Protein G purified | |
RUO | |
Primary | |
PDGF Receptor beta (42G12) | |
Rat, Mouse, Human | |
Purified |
In situ PLA, KnockDown, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
42G12 | |
Western Blot 1:100-1:2000, Immunohistochemistry 20 ug/mL, Immunohistochemistry-Paraffin 20 ug/mL, Proximity Ligation Assay, KnockDown Validated | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
Mouse | |
125 kDa | |
0.025 mg | |
Protein Kinase | |
5159 | |
PBS, 50% glycerol, pH 7.2 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction