Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Mouse anti-Human, Mouse, Rat, Clone: 42G12, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
$176.85 - $436.00
Specifications
Antigen | PDGF R beta |
---|---|
Clone | 42G12 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), In situ PLA, KnockDown |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Proximity Ligation Assay, Knockdown Validated.Specifications
PDGF R beta | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), In situ PLA, KnockDown | |
Unconjugated | |
Mouse | |
Protein Kinase | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
PDGFRB | |
IgG1 | |
Protein G purified | |
Store at -20C. Avoid freeze-thaw cycles. | |
125 kDa |
42G12 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
5159 | |
Recombinant human PDGF Receptor beta Protein | |
Primary | |
PBS, 50% glycerol, pH 7.2 | |
PDGF Receptor beta (42G12) |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title