Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Rat anti-Mouse, Clone: RM0005-4C54, Novus Biologicals™
Rat Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | PDGF R beta |
---|---|
Clone | RM0005-4C54 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-200, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11061021
|
Novus Biologicals
NB11061021 |
0.1 mg |
Each for $493.00
|
|
NB018000
|
Novus Biologicals
NB110610210.025MG |
0.025 mg |
Each for $177.16
|
|
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PDGF R beta | |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
Protein Kinase | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
PDGFRB | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
RM0005-4C54 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
5159 | |
IgG2 | |
Protein G purified | |
PDGF receptor beta (RM0005-4C54) |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title