Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFR alpha, Human anti-Human, Clone: Hu136, R&D Systems™

Human Monoclonal Antibody

$131.70 - $334.00

Specifications

Antigen PDGFR alpha
Clone Hu136
Immunogen Human PDGF R alpha
Host Species Human
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB10352100
View Documents
R&D Systems
MAB10352100
100 ug
Each for $334.00
Add to cart
 
MAB10352SP
View Documents
R&D Systems
MAB10352SP
25 ug
Each for $131.70
Add to cart
 
Description

Description

PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human Samples. It is validated for Flow Cytometry, CyTOF.
Specifications

Specifications

PDGFR alpha
Human PDGF R alpha
Primary
Human
Unconjugated
5156
Purified
Flow Cytometry, CyTOF
RUO
Hu136
Human
Monoclonal
IgG1
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit