Learn More
MilliporeSigma™ PDGFR Beta, Rabbit, Unlabeled, Polyclonal,
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ 06498I
Catalog No. 06498IMI
Add to cart
Description
Description
Platelet-derived growth factor receptor beta (EC 2.7.10.1; UniProt P09619; also known as Beta platelet-derived growth factor receptor, Beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, PDGF-R-beta, PDGFR-1, PDGFR-beta, Platelet-derived growth factor receptor 1) is encoded by the PDGFRB (also known as IBGC4, IMF1, PDGFR, PDGFR1) gene (Gene ID 5159) in human. PDGFR-1 is a tyrosine-protein kinase that acts as a cell-surface receptor for homodimeric PDGFB and PDGFD and for heterodimers formed by PDGFA and PDGFB. PDGF receptors consist of extracellular domains with five immunoglobulin (Ig) - like domains and intracellular parts with kinase domains, which contain characteristic inserts of about 100 amino acid residues without homology to kinases. Ligand binding occurs mainly to Ig-like domains 2 and 3, which causes dimerization of the receptor, which is further stabilized by direct receptor-receptor interactions involving Ig-like domain 4. In the absence of a bound ligand, receptor is present in an inactive conformation. However, upon ligand binding it undergoes dimerization and autophosphorylation on tyrosine residues. PDGFR-1m plays an essential role in the regulation of embryonic development, cell proliferation, survival, differentiation, chemotaxis, and migration. It is also considered to be essential for endothelial cell proliferation, migration and recruitment of pericytes and smooth muscle cells to form blood vessels. PDGFR-1 phosphorylates phospholipase C-gamma 1 (PLCG1), PIK3R1, PTPN11, RASA1/GAP, CBL, SHC1 and NCK1. Activation of PLCG1 leads to the production of diacylglycerol and IP3 that are involved in calcium mobilization and activation of PKC. Ref.: Heldin C-H (2013). Cell Commun. Signaling 11:97.C1
Specifications
Specifications
PDGFR Beta
Please refer to lot specific datasheet.
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
P09619
Rabbit
Affinity Purified
RUO
Stable for 1 year at 2°-8°C from date of receipt.
Polyclonal
This polyclonal antibody targets a cytoplasmic sequence C-terminal to the PDGFR kinase domain.
Immunoprecipitation, Western Blot
Unconjugated
PDGFRB, IBGC4, IMF1, PDGFR, PDGFR1
Affinity Purified
KLH-conjugated linear peptide corresponding to a cytoplasmic sequence C-terminal to the kinase domain of human PDGFR .
100μg
Signaling
Primary
NP_002600
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.