Learn More
Selleck Chemical LLCSupplier Diversity Partner PDGFR inhibitor 1 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S87215MG
PDGFR inhibitor 1 is an orally bioavailable switch pocket control inhibitor of wild-type and mutated forms of KIT and PDGFR with potential antineoplastic activity. It also inhibits several other kinases, including VEGFR2, TIE2, PDGFR-beta and CSF1R, thereby further inhibiting tumor cell growth.
Catalog No. 50-194-8383