Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ PDGFRα, Rabbit Polyclonal Antibody

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ PDGFRα, Rabbit Polyclonal Antibody.

$207.50 - $961.50

Specifications

Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status IVD
View More Specs

Products 5
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
RB9027P
View Documents
Thermo Scientific™
RB9027P
Concentrated 1mL Each for $961.50
Add to cart
 
RB9027P0
View Documents
Thermo Scientific™
RB9027P0
Concentrated 100μL Each for $207.50
Add to cart
 
RB9027P1
View Documents
Thermo Scientific™
RB9027P1
Concentrated 500μL Each for $739.50
Add to cart
 
RB9027R7
View Documents
Thermo Scientific™
RB9027R7
Prediluted 7mL Each for $533.00
Add to cart
 
RB9027R1
View Documents
Thermo Scientific™
RB9027R1
Prediluted 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ PDGFRα, Rabbit Polyclonal Antibody.

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, alpha; and beta; coded by two different genes. PDGF A receptors are expressed in oligodendrocyte progenitor cells and PDGF B receptors are expressed on neurons. This antibody is specific for PDGFR, alpha; and does not react with PDGFR, beta;.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from C-terminal of human PDGFRα

Molecular Weight: 190kDa

Positive Control: Skin and ovarian CA

Cellular Localization: Cytoplasmic and cell membrane This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG
Polyclonal
Rabbit
Growth Factor
A synthetic peptide derived from C-terminal of human PDGFR alpha
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit