Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRA Mouse anti-Human, APC, Clone: 16A1, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A15718

Catalog No. 50-113-7074


Add to cart

Description

Description

PDGFRA is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.
Specifications

Specifications

PDGFRA
Monoclonal
APC
PDGFRA
Liquid
PDGFRA
Human CD104a
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
16A1
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P16234
PDGF Receptor alpha, MGC74795, PDGFR2, RHEPDGFRA, CD140 antigen-like family member A, CD140a antigen, PDGF-R-alpha, PDGFRA/BCR fusion, pdgfr-a
Mouse
IgG1
100 tests
4° C
Primary
5156
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit