Learn More
PDGFRA Mouse anti-Human, Biotin, Clone: 16A1, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15732
Catalog No. 50-113-7083
Add to cart
Description
Description
PDGFRA is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.
Specifications
Specifications
PDGFRA
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
P16234
PDGF Receptor alpha, MGC74795, PDGFR2, RHEPDGFRA, CD140 antigen-like family member A, CD140a antigen, PDGF-R-alpha, PDGFRA/BCR fusion, pdgfr-a
Mouse
IgG1
100 μg
4° C
Primary
5156
Flow Cytometry
16A1
Biotin
PDGFRA
Liquid
PDGFRA
Human CD104a
Purified
RUO
Antibody
Monoclonal
Human